Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ang | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161731.2 | NP_001155203.1 | 2 | 481 | ||
NM_007447.3 | NP_031473.1 | 2 | 778 | |||
GenBank mRNA | AK159966.1 | - | 2 | 779 | ||
AK171251.1 | - | 2 | 409 | |||
BC055355.1 | - | 2 | 365 | 5136641 | ||
BF721354.1 | - | 2 | 354 |