Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF004670.1 | - | 2 | 227 | ||
AF093852.1 | - | 2 | 258 | |||
AK030413.1 | - | 2 | 260 | |||
AK157820.1 | - | 2 | 277 | |||
AK161710.1 | - | 2 | 260 | |||
AK168223.1 | - | 2 | 231 | |||
AK172888.1 | - | 2 | 220 | |||
BC013489.1 | - | 2 | 219 | 4215591 | ||
BC061181.1 | - | 2 | 259 | 30309446 | ||
Y12883.1 | - | 2 | 277 |