Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077264.1 | NP_001070732.1 | 3 | 403 | ||
NM_007458.2 | NP_031484.1 | 3 | 403 | |||
GenBank mRNA | AK135424.1 | - | 3 | 332 | ||
AK155515.1 | - | 3 | 339 | |||
AK208311.1 | - | 2 | 190 | |||
AK212972.1 | - | 3 | 308 | |||
BC031433.1 | - | 3 | 353 | 4481643 | ||
CA327188.1 | - | 3 | 409 |