Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF084575.1 | - | 6 | 603 | ||
AK002565.1 | - | 6 | 629 | |||
AK137489.1 | - | 6 | 624 | |||
AK152100.1 | - | 6 | 635 | |||
BC012656.1 | - | 6 | 614 | 4187607 | ||
BC108327.1 | - | 6 | 622 | 30713247 | ||
BC131985.1 | - | 6 | 604 | 40130262 | ||
BC145234.1 | - | 7 | 645 |