Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007468.2 | NP_031494.2 | 2 | 467 | ||
GenBank mRNA | AK004856.1 | - | 2 | 250 | ||
AK161535.1 | - | 2 | 250 | |||
AK168687.1 | - | 2 | 251 | |||
BC010769.1 | - | 2 | 260 | 4240796 | ||
BC050149.1 | - | 2 | 225 | 30013747 | ||
BF660098.1 | - | 2 | 479 | |||
M64248.1 | - | 2 | 249 | |||
M64249.1 | - | 2 | 249 | |||
M64250.1 | - | 2 | 249 |