Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019734.2 | NP_062708.1 | 10 | 830 | ||
GenBank mRNA | AF157500.1 | - | 10 | 760 | ||
AF352179.1 | - | 10 | 772 | |||
AK075666.1 | - | 10 | 745 | |||
AK075919.1 | - | 10 | 784 | |||
AK077721.1 | - | 10 | 783 | |||
AK090314.1 | - | 10 | 780 | |||
AK150567.1 | - | 10 | 742 | |||
AK151208.1 | - | 10 | 831 | |||
AK151312.1 | - | 10 | 742 | |||
AK151478.1 | - | 10 | 821 | |||
AK152305.1 | - | 10 | 782 | |||
AK152811.1 | - | 10 | 742 | |||
AK152920.1 | - | 10 | 790 | |||
AK153101.1 | - | 10 | 790 | |||
AK153297.1 | - | 10 | 744 | |||
AK159556.1 | - | 10 | 752 | |||
AK166757.1 | - | 10 | 762 | |||
BC003204.1 | - | 10 | 750 | 3587042 |