Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ass1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007494.3 | NP_031520.1 | 3 | 223 | ||
GenBank mRNA | AK146635.1 | - | 3 | 192 | ||
AK149489.1 | - | 3 | 195 | |||
BC002074.1 | - | 3 | 223 | 3491910 | ||
BC087556.1 | - | 2 | 152 | 6478590 | ||
M31690.1 | - | 3 | 193 |