Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007496.2 | NP_031522.2 | 9 | 9803 | ||
GenBank mRNA | AK220555.1 | - | 1 | 1263 | ||
BC060729.1 | - | 1 | 3022 | |||
BC082769.1 | - | 1 | 738 | |||
D26046.1 | - | 9 | 9800 |