Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000561.3 | NP_000552.2 | 5 | 419 | ||
NM_146421.2 | NP_666533.1 | 5 | 419 | |||
XM_005270782.4 | XP_005270839.1 | 5 | 437 | |||
GenBank mRNA | AY510272.1 | - | 5 | 341 | ||
AY532926.1 | - | 5 | 341 | |||
AY532927.1 | - | 5 | 341 | |||
BC024005.2 | - | 5 | 375 | 4823313 | ||
BC036805.1 | - | 5 | 356 | |||
BQ436476.1 | - | 5 | 405 | |||
CR541868.1 | - | 5 | 341 | |||
DB459176.1 | - | 5 | 421 | |||
J03817.1 | - | 5 | 356 | |||
KJ534856.1 | - | 5 | 341 | |||
KJ534857.1 | - | 5 | 341 | |||
KJ534858.1 | - | 5 | 341 | |||
KJ534998.1 | - | 4 | 259 | |||
KJ535021.1 | - | 4 | 392 | |||
X08020.1 | - | 5 | 365 |