Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000848.3 | NP_000839.1 | 7 | 596 | ||
NM_001142368.1 | NP_001135840.1 | 7 | 596 | |||
GenBank mRNA | AK299482.1 | - | 7 | 725 | ||
AK309796.1 | - | 7 | 513 | |||
BC105038.1 | - | 7 | 527 | 8144041 | ||
BC105066.1 | - | 7 | 527 | 8144069 | ||
BC110380.1 | - | 7 | 549 | 5760414 | ||
BM924181.1 | - | 7 | 562 | |||
BT019947.1 | - | 7 | 502 | |||
M63509.1 | - | 7 | 518 |
Gene Symbol: GSTM2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002932.2 | 1 | 535 | |||
GenBank mRNA | AY532924.1 | - | 1 | 502 | ||
AY532925.1 | - | 1 | 502 |