Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009724.2 | NP_033854.1 | 2 | 166 | ||
GenBank mRNA | AK008180.1 | - | 2 | 165 | ||
AK008686.1 | - | 2 | 166 | |||
AK009499.1 | - | 2 | 166 | |||
AK009670.1 | - | 2 | 171 | |||
AK009832.1 | - | 2 | 168 | |||
AK009994.1 | - | 2 | 165 | |||
AK010003.1 | - | 2 | 165 | |||
AK010030.1 | - | 2 | 166 | |||
AK010049.1 | - | 2 | 213 | |||
BC120705.1 | - | 2 | 165 | 40129628 | ||
BC125292.1 | - | 2 | 165 | 40129807 |