Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077593.1 | - | 8 | 1339 | ||
AK141558.1 | - | 8 | 1370 | |||
AK165931.1 | - | 8 | 1336 | |||
BC050195.1 | - | 8 | 1315 | 30010078 | ||
U70859.1 | - | 9 | 1473 |