Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312660.1 | NP_001299589.1 | 4 | 575 | ||
NM_145870.2 | NP_665877.1 | 3 | 389 | |||
NM_145871.2 | NP_665878.2 | 3 | 389 | |||
XM_011536670.2 | XP_011534972.1 | 4 | 631 | |||
XM_011536671.2 | XP_011534973.1 | 3 | 299 | |||
XM_017021243.1 | XP_016876732.1 | 3 | 301 | |||
GenBank mRNA | AB209360.1 | - | 1 | 4156 | ||
AJ001838.1 | - | 3 | 210 | |||
AK309054.1 | - | 3 | 237 | |||
AK315154.1 | - | 4 | 466 | |||
AK315609.1 | - | 3 | 233 | |||
BC001453.1 | - | 3 | 265 | 3139094 | ||
BE958109.1 | - | 196 | ||||
BG330365.1 | - | 3 | 251 | |||
BG772450.1 | - | 3 | 396 | |||
BI559916.1 | - | 4 | 484 | |||
BI829992.1 | - | 4 | 455 | |||
CR456987.1 | - | 2 | 107 | |||
U86529.1 | - | 3 | 210 |