Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291061.1 | NP_001277990.1 | 3 | 674 | ||
NM_009751.2 | NP_033881.2 | 3 | 674 | |||
GenBank mRNA | AB003147.1 | - | 3 | 444 | ||
AK053766.1 | - | 3 | 445 | |||
AK143180.1 | - | 3 | 445 | |||
BC057580.1 | - | 3 | 674 | 6412634 |