Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009752.2 | NP_033882.1 | 6 | 748 | ||
GenBank mRNA | AK034049.1 | - | 6 | 727 | ||
AK034966.1 | - | 549 | ||||
AK042069.1 | - | 6 | 718 | |||
AK048460.1 | - | 6 | 679 | |||
AK076517.1 | - | 6 | 719 | |||
AK153218.1 | - | 6 | 731 | |||
AK166473.1 | - | 6 | 733 | |||
AK167271.1 | - | 4 | 358 | |||
AK171593.1 | - | 6 | 751 | |||
AK171760.1 | - | 5 | 549 | |||
BC003998.1 | - | 4 | 319 | |||
BC028875.1 | - | 6 | 668 | 4240285 | ||
M57734.1 | - | 6 | 707 |