Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009754.3 | NP_033884.1 | 3, 4 | 463 | ||
NM_207680.2 | NP_997563.1 | 3, 4 | 721 | |||
NM_207681.2 | NP_997564.1 | 4, 5 | 553 | |||
GenBank mRNA | AF032459.1 | - | 2, 3 | 493 | ||
AF032460.1 | - | 3, 4 | 325 | |||
AF032461.1 | - | 2, 3 | 235 | |||
AK011490.1 | - | 3, 4 | 710 | |||
AK050192.1 | - | 4, 5 | 542 | |||
AK082613.1 | - | 3, 4 | 717 | |||
AK150464.1 | - | 3, 4 | 464 | |||
AK152472.1 | - | 3, 4 | 464 | |||
AK153542.1 | - | 3, 4 | 464 | |||
BC019556.1 | - | 3, 4 | 715 | 4459720 |