Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007552.4 | NP_031578.2 | 9 | 1087 | ||
GenBank mRNA | AK145437.1 | - | 9 | 742 | ||
BC053708.1 | - | 9 | 713 | 30074540 | ||
BC056384.1 | - | 8 | 617 | 6854104 | ||
DQ337175.1 | - | 8 | 616 | |||
M64279.1 | - | 9 | 1087 |