Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019506.4 | NP_062379.3 | 2 | 764 | ||
GenBank mRNA | AK147029.1 | - | 2 | 765 | ||
BC103625.1 | - | 2 | 733 | 40047137 | ||
BC103679.1 | - | 2 | 733 | 40047139 | ||
BC103680.1 | - | 2 | 733 | 40047138 | ||
BC103681.1 | - | 2 | 733 | 40047136 |