Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160127.1 | NP_001153599.1 | 9 | 1372 | ||
NM_009762.2 | NP_033892.2 | 8 | 1333 | |||
GenBank mRNA | AK076266.1 | - | 8 | 1298 | ||
AK142153.1 | - | 8 | 1301 | |||
AK142252.1 | - | 9 | 1350 | |||
BC076601.1 | - | 9 | 1350 | 30609878 | ||
U76371.1 | - | 1477 | ||||
U76373.2 | - | 9 | 1344 | |||
U76374.2 | - | 8 | 1305 |