Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081001.2 | NP_001074470.1 | 10 | 1846 | ||
NM_009765.3 | NP_033895.2 | 10 | 1652 | |||
GenBank mRNA | AK145017.1 | - | 10 | 1813 | ||
AK148272.1 | - | 10 | 1649 | |||
AK164598.1 | - | 8 | 2158 | |||
U65594.1 | - | 10 | 1629 | |||
U82270.1 | - | 9 | 1338 | |||
U89652.1 | - | 9 | 1376 |