Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081001.2 | NP_001074470.1 | 11 | 4990 | ||
NM_009765.3 | NP_033895.2 | 11 | 4796 | |||
GenBank mRNA | U65594.1 | - | 11 | 4773 | ||
U82270.1 | - | 10 | 4482 | |||
U89652.1 | - | 10 | 4520 |