Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252533.1 | NP_001239462.1 | 10 | 1182 | ||
NM_023116.4 | NP_075605.1 | 11 | 1313 | |||
GenBank mRNA | AK034054.1 | - | 11 | 1314 | ||
AM259383.2 | - | 10 | 1182 | |||
BC109156.1 | - | 11 | 1245 | 40047524 | ||
BC109157.1 | - | 11 | 1245 | 40047531 | ||
KJ789960.1 | - | 10 | 901 | |||
L20343.1 | - | 10 | 1092 |