Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109761.2 | NP_001103231.1 | 18 | 1962 | ||
NM_001177799.1 | NP_001171270.1 | 18 | 2229 | |||
NM_007601.3 | NP_031627.2 | 21 | 2505 | |||
GenBank mRNA | AF091998.1 | - | 18 | 1926 | ||
AF127766.2 | - | 18 | 1949 | |||
AK049994.1 | - | 8 | 391 | |||
AK079673.1 | - | 3 | 181 | |||
AK088613.1 | - | 6 | 716 | |||
BC090661.1 | - | 18 | 1962 | 30681055 | ||
BC139790.1 | - | 21 | 2225 | 40047690 | ||
X92523.1 | - | 21 | 2225 |