Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007606.3 | NP_031632.2 | 6 | 603 | ||
GenBank mRNA | AK002349.1 | - | 6 | 630 | ||
AK003671.1 | - | 6 | 633 | |||
M27796.1 | - | 6 | 652 |