Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007611.2 | NP_031637.1 | 6, 7 | 872 | ||
GenBank mRNA | AK154978.1 | - | 6, 7 | 752 | ||
BC005428.1 | - | 6, 7 | 729 | 3600155 | ||
CT010348.1 | - | 5, 6 | 683 | |||
D86353.1 | - | 6, 7 | 735 | |||
Y13088.1 | - | 6, 7 | 872 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U67321.1 | - | 1267 | Chr. - - [-] on Build |