Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080126.1 | NP_001073595.1 | 8 | 1455 | ||
NM_001277926.1 | NP_001264855.1 | 9 | 1571 | |||
NM_009812.2 | NP_033942.1 | 8 | 1628 | |||
GenBank mRNA | AF067834.1 | - | 8 | 1508 | ||
AJ000641.1 | - | 7 | 1074 | |||
AJ007749.1 | - | 8 | 1281 | |||
AK146758.1 | - | 8 | 1628 | |||
AK153340.1 | - | 8 | 1337 | |||
AK156135.1 | - | 8 | 1344 | |||
AK168812.1 | - | 8 | 1628 | |||
BC006737.1 | - | 8 | 1570 | 3964119 | ||
BC049955.1 | - | 8 | 1462 | 4208824 | ||
CT010166.1 | - | 7 | 1235 |