Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161457.1 | NP_001154929.1 | 3 | 492 | ||
NM_001161458.1 | NP_001154930.1 | 3 | 492 | |||
NM_022309.4 | NP_071704.3 | 3 | 492 | |||
GenBank mRNA | AK089094.1 | - | 3 | 493 | ||
AK134449.1 | - | 2 | 60 | |||
BC006763.1 | - | 3 | 260 | 3481800 | ||
BC026749.1 | - | 3 | 302 | 4016060 | ||
BC040752.1 | - | 3 | 291 | 4016060 | ||
D14570.1 | - | 3 | 422 | |||
D14571.1 | - | 3 | 422 | |||
D14572.1 | - | 3 | 422 | |||
L03279.1 | - | 3 | 271 | |||
L03306.1 | - | 190 |