Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000856.5 | NP_000847.2 | 9, 10 | 2249 | ||
NM_001130682.2 | NP_001124154.1 | 8, 9 | 2155 | |||
NM_001130683.3 | NP_001124155.1 | 8, 9 | 2123 | |||
NM_001130684.2 | NP_001124156.1 | 7, 8 | 2095 | |||
NM_001130685.2 | NP_001124157.1 | 8, 9 | 1944 | |||
NM_001130687.2 | NP_001124159.1 | 8, 9 | 2155 | |||
NM_001256449.1 | NP_001243378.1 | 8, 9 | 2086 | |||
XM_005262955.2 | XP_005263012.1 | 8, 9 | 2117 | |||
XM_005262956.2 | XP_005263013.1 | 9, 10 | 1932 | |||
XM_005262957.2 | XP_005263014.1 | 9, 10 | 2037 | |||
XM_006714196.2 | XP_006714259.1 | 8, 9 | 2104 | |||
XM_006714197.2 | XP_006714260.1 | 8, 9 | 1879 | |||
XM_006714198.2 | XP_006714261.1 | 8, 9 | 1764 | |||
XM_011531900.2 | XP_011530202.1 | 8, 9 | 1843 | |||
GenBank mRNA | AK096588.1 | - | 8, 9 | 2086 | ||
AK226125.1 | - | 8, 9 | 1812 | |||
AK309950.1 | - | 8, 9 | 2017 | |||
AK312952.1 | - | 8, 9 | 2055 | |||
BC012627.1 | - | 5, 6 | 715 | |||
BC028384.2 | - | 7, 8 | 2095 | 4792661 | ||
BC095395.1 | - | 8, 9 | 2046 | |||
BX647200.1 | - | 8, 9 | 1922 | |||
BX649180.1 | - | 8, 9 | 1876 | |||
U58855.1 | - | 6, 7 | 1730 | |||
X66534.1 | - | 9, 10 | 2226 | |||
Y15723.1 | - | 9, 10 | 2236 |