Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007625.2 | NP_031651.2 | 5 | 1016 | ||
GenBank mRNA | AK155152.1 | - | 1 | 114 | ||
AK155512.1 | - | 1 | 114 | |||
BC018304.1 | - | 5 | 1047 | |||
BC115843.1 | - | 2 | 693 | |||
BC115844.1 | - | 1 | 908 | |||
BC117801.1 | - | 5 | 890 | 40092282 | ||
BC117802.1 | - | 5 | 921 | 40092283 | ||
U63387.1 | - | 5 | 928 |