Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076789.1 | NP_001070257.1 | 5 | 620 | ||
NM_001110216.1 | NP_001103686.1 | 6 | 721 | |||
NM_007626.3 | NP_031652.1 | 5 | 621 | |||
GenBank mRNA | AK008792.1 | - | 5 | 656 | ||
AK018349.1 | - | 5 | 567 | |||
AK030366.1 | - | 5 | 621 | |||
AK030442.1 | - | 6 | 674 | |||
AK032975.1 | - | 5 | 577 | |||
BC004707.1 | - | 5 | 620 | 3499489 | ||
X99641.1 | - | 5 | 553 |