Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007634.4 | NP_031660.3 | 15 | 1717 | ||
GenBank mRNA | AK030298.1 | - | 1656 | |||
AK032961.1 | - | 15 | 1643 | |||
AK082485.1 | - | 15 | 1685 | |||
AK085183.1 | - | 15 | 1653 | |||
AK159594.1 | - | 15 | 1718 | |||
AK169263.1 | - | 15 | 1668 | |||
AK214285.1 | - | 4 | 268 |