Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009838.2 | NP_033968.1 | 1434 | |||
GenBank mRNA | AK077916.1 | - | 1388 | |||
AK134428.1 | - | 1387 | ||||
AK145522.1 | - | 1394 | ||||
AK147130.1 | - | 1431 | ||||
AK149757.1 | - | 1793 | ||||
AK150977.1 | - | 1388 | ||||
AK159354.1 | - | 1388 | ||||
AK159785.1 | - | 1408 | ||||
AK159975.1 | - | 1434 | ||||
AK160067.1 | - | 1388 | ||||
AK164312.1 | - | 1388 | ||||
AK165612.1 | - | 1390 | ||||
AK166749.1 | - | 1388 | ||||
AK166795.1 | - | 1418 | ||||
AK166870.1 | - | 1386 | ||||
AK166887.1 | - | 1387 | ||||
AK166929.1 | - | 1386 | ||||
AK167007.1 | - | 1416 | ||||
AK167076.1 | - | 1385 | ||||
AK167082.1 | - | 1384 | ||||
AK167164.1 | - | 1393 | ||||
AK167180.1 | - | 1420 | ||||
AK167307.1 | - | 1388 | ||||
AK167669.1 | - | 1417 | ||||
AK167851.1 | - | 1389 | ||||
AK167994.1 | - | 1383 | ||||
AK168065.1 | - | 1383 | ||||
AK168940.1 | - | 1417 | ||||
AK169033.1 | - | 1367 | ||||
AK190808.1 | - | 126 | ||||
AK201996.1 | - | 126 | ||||
BC018459.1 | - | 998 | 3597455 | |||
BC055043.1 | - | 1396 | ||||
BC069917.1 | - | 1389 | 6394714 | |||
BC094352.1 | - | 1360 | 6809215 | |||
Z31557.1 | - | 1405 |