Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009840.3 | NP_033970.3 | 11 | 1244 | ||
GenBank mRNA | AK010882.1 | - | 11 | 1245 | ||
AK014005.1 | - | 4 | 310 | |||
AK075711.1 | - | 11 | 1225 | |||
AK075880.1 | - | 11 | 1233 | |||
AK145661.1 | - | 11 | 1228 | |||
AK145754.1 | - | 11 | 1227 | |||
AK145918.1 | - | 11 | 1246 | |||
AK147038.1 | - | 11 | 1247 | |||
AK159969.1 | - | 11 | 1235 | |||
AK168465.1 | - | 11 | 1217 | |||
AK172867.1 | - | 11 | 1215 | |||
AK218945.1 | - | 4 | 271 | |||
BC009007.1 | - | 11 | 1202 | 3582639 | ||
Z37164.1 | - | 11 | 1248 |