Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161587.1 | NP_001155059.1 | 6 | 1011 | ||
NM_002103.4 | NP_002094.2 | 7 | 1203 | |||
NR_027763.1 | 6 | 1021 | ||||
GenBank mRNA | AK223579.1 | - | 7 | 1159 | ||
AK291672.1 | - | 7 | 1159 | |||
AK294349.1 | - | 2 | 119 | |||
AK300369.1 | - | 7 | 991 | |||
AK302738.1 | - | 6 | 977 | |||
AK303712.1 | - | 5 | 859 | |||
BC002617.2 | - | 7 | 1142 | 3143019 | ||
BC003182.1 | - | 6 | 968 | 2958297 | ||
BC007688.2 | - | 7 | 1129 | 3953692 | ||
J04501.1 | - | 7 | 1122 | |||
U32573.1 | - | 7 | 1104 |