Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007658.3 | NP_031684.3 | 6 | 890 | ||
GenBank mRNA | AK077609.1 | - | 6 | 507 | ||
AK077991.1 | - | 6 | 893 | |||
AK142334.1 | - | 5 | 1165 | |||
AK213270.1 | - | 2 | 101 | |||
BC046296.1 | - | 6 | 509 | 6486343 |