Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007664.4 | NP_031690.3 | 8 | 1687 | ||
GenBank mRNA | AB008811.1 | - | 8 | 1364 | ||
AK032826.1 | - | 8 | 1127 | |||
AK146981.1 | - | 8 | 1884 | |||
BC022107.1 | - | 8 | 1118 | 5100313 | ||
M31131.1 | - | 8 | 1364 |