Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA016357.1 | - | 2, 3 | 262 | ||
AF012709.1 | - | 2, 3 | 318 | |||
AK010718.1 | - | 2, 3 | 280 | |||
AK011399.1 | - | 2, 3 | 263 | |||
AK040313.1 | - | 3, 4 | 445 | |||
AK041138.1 | - | 3, 4 | 438 | |||
AK053763.1 | - | 2, 3 | 287 | |||
AK089033.1 | - | 2, 3 | 291 | |||
BC011038.1 | - | 2, 3 | 298 | 4018429 | ||
BC012280.1 | - | 4, 5 | 391 |