Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002104.2 | NP_002095.1 | 4 | 488 | ||
GenBank mRNA | AK312074.1 | - | 4 | 511 | ||
BC035802.1 | - | 4 | 488 | 5748080 | ||
U26174.1 | - | 4 | 484 | |||
U35237.1 | - | 4 | 444 |