Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163432.1 | NP_001156904.1 | 13 | 1657 | ||
NM_007712.3 | NP_031738.2 | 13 | 1660 | |||
GenBank mRNA | AF033564.1 | - | 13 | 1382 | ||
AK148377.1 | - | 11 | 2688 | |||
AK161259.1 | - | 9 | 1832 | |||
BC015080.1 | - | 13 | 1645 | 3995512 |