Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002106.3 | NP_002097.1 | 3 | 286 | ||
GenBank mRNA | AK056803.1 | - | 2 | 562 | ||
AK315413.1 | - | 3 | 204 | |||
AY597416.1 | - | 3 | 223 | |||
BC018002.1 | - | 3 | 206 | 4271353 | ||
BC020936.1 | - | 3 | 206 | 4281642 | ||
BC103743.1 | - | 3 | 321 | 6175124 | ||
CD685635.1 | - | 3 | 319 | |||
CR457415.1 | - | 2 | 101 | |||
M37583.1 | - | 3 | 207 | |||
X52317.1 | - | 3 | 207 |