Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007742.3 | NP_031768.2 | 2, 3 | 369 | ||
GenBank mRNA | AK132180.1 | - | 2, 3 | 385 | ||
AK158011.1 | - | 2, 3 | 385 | |||
AK159283.1 | - | 2, 3 | 387 | |||
AK159285.1 | - | 2, 3 | 389 | |||
BC050014.1 | - | 2, 3 | 369 | 4225038 | ||
BC059281.1 | - | 2, 3 | 379 | 3992363 | ||
U08020.1 | - | 2, 3 | 270 |