Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 46 | 3193 | ||
GenBank mRNA | AK075707.1 | - | 24 | 1765 | ||
AK132496.1 | - | 46 | 3189 | |||
AK159120.1 | - | 6 | 394 | |||
AK159212.1 | - | 46 | 3195 | |||
AK159408.1 | - | 46 | 3190 | |||
AK159578.1 | - | 46 | 3192 | |||
AK159958.1 | - | 25 | 1813 | |||
AK160098.1 | - | 46 | 3194 | |||
AK160787.1 | - | 46 | 3189 | |||
AK160788.1 | - | 46 | 3188 | |||
AK160790.1 | - | 46 | 3189 | |||
AK160814.1 | - | 38 | 2658 | |||
AK160946.1 | - | 46 | 3192 | |||
AK161017.1 | - | 46 | 3194 | |||
AK161605.1 | - | 46 | 3189 | |||
AK164628.1 | - | 25 | 1812 | |||
AK164808.1 | - | 46 | 3189 | |||
BC007158.1 | - | 46 | 3173 | 3487477 | ||
BC012438.1 | - | 7 | 633 | |||
BC042503.1 | - | 46 | 3181 | 3586272 | ||
X58251.1 | - | 46 | 3129 |