Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013496.3 | NP_038524.1 | 3 | 418 | ||
GenBank mRNA | AK045283.1 | - | 3 | 360 | ||
AK144397.1 | - | 3 | 370 | |||
M31552.1 | - | 2 | 71 | |||
X15481.1 | - | 3 | 344 | |||
X15789.1 | - | 3 | 360 |