Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040427.1 | NP_001035517.1 | 10 | 1564 | ||
NM_001286249.1 | NP_001273178.1 | 8 | 1359 | |||
NM_005326.4 | NP_005317.2 | 9 | 1468 | |||
XM_011522469.2 | XP_011520771.1 | 9 | 1172 | |||
XM_011522470.2 | XP_011520772.1 | 8 | 1061 | |||
XM_017023195.1 | XP_016878684.1 | 9 | 1097 | |||
GenBank mRNA | AK299173.1 | - | 9 | 1099 | ||
BC000840.1 | - | 9 | 1088 | 3457214 | ||
BC002627.2 | - | 9 | 1417 | 3608098 | ||
X90999.1 | - | 8 | 953 |