Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037726.1 | NP_001032815.1 | 5, 6 | 783 | ||
NM_009952.2 | NP_034082.1 | 7, 8 | 947 | |||
NM_133828.2 | NP_598589.2 | 6, 7 | 905 | |||
GenBank mRNA | AF448507.1 | - | 7, 8 | 823 | ||
AF448508.1 | - | 1475 | ||||
AK044992.1 | - | 6, 7 | 720 | |||
AK044993.1 | - | 6, 7 | 720 | |||
AK079223.1 | - | 6, 7 | 901 | |||
BB831364.1 | - | 2, 3 | 277 | |||
BC021649.1 | - | 6, 7 | 838 | 4017170 | ||
M95106.1 | - | 7, 8 | 857 | |||
X92497.1 | - | 5, 6 | 760 |