Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013497.3 | NP_038525.2 | 2, 3 | 480 | ||
GenBank mRNA | AK090077.1 | - | 2, 3 | 450 | ||
AK132785.1 | - | 2, 3 | 490 | |||
AK144208.1 | - | 2, 3 | 483 | |||
BC002094.1 | - | 2, 3 | 446 | 3490360 |