Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025432.1 | NP_001020603.1 | 19, 20 | 3876 | ||
GenBank mRNA | BC072594.1 | - | 19, 20 | 3862 | ||
S66385.1 | - | 19, 20 | 3695 |