Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040427.1 | NP_001035517.1 | 9 | 1301 | ||
NM_001286249.1 | NP_001273178.1 | 7 | 1096 | |||
NM_005326.4 | NP_005317.2 | 8 | 1205 | |||
XM_011522469.2 | XP_011520771.1 | 8 | 845 | |||
XM_011522470.2 | XP_011520772.1 | 7 | 734 | |||
XM_017023195.1 | XP_016878684.1 | 8 | 770 | |||
GenBank mRNA | AK054943.1 | - | 2 | 1168 | ||
AK290155.1 | - | 8 | 770 | |||
AK298174.1 | - | 7 | 736 | |||
AK299173.1 | - | 8 | 836 | |||
AK299993.1 | - | 8 | 836 | |||
BC000840.1 | - | 8 | 825 | 3457214 | ||
BC002627.2 | - | 8 | 1154 | 3608098 | ||
X90999.1 | - | 7 | 690 |