Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007763.3 | NP_031789.1 | 2 | 194 | ||
GenBank mRNA | AA266159.1 | - | 2 | 194 | ||
AI323004.1 | - | 2 | 142 | |||
AK003075.1 | - | 2 | 156 | |||
AK008269.1 | - | 2 | 156 | |||
AK012068.2 | - | 2 | 151 | |||
AK088267.1 | - | 2 | 156 | |||
AK168305.1 | - | 2 | 155 | |||
BC030406.1 | - | 1 | 1264 | |||
BC031922.1 | - | 2 | 139 | 4206322 | ||
BC058606.1 | - | 1 | 1238 | |||
BC064074.1 | - | 2 | 143 | 948562 | ||
BX511593.1 | - | 2 | 140 | |||
DT908028.1 | - | 2 | 194 |