Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009968.3 | NP_034098.1 | 3 | 826 | ||
GenBank mRNA | AK098082.1 | - | 3 | 310 | ||
AK133820.1 | - | 3 | 827 | |||
AK134281.1 | - | 3 | 311 | |||
BC043076.1 | - | 3 | 287 | 6404996 | ||
S70056.1 | - | 3 | 338 |